Order Entry
Norway
ContactUsLinkComponent
 
 
3,4,5-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
3,4,5-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    168.0 to 173.0 deg-C
  • Solubility in Methanol:
    very faint turbidity