

3-Nitrobenzoic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:121-92-6
:
Formula:C₇H₅NO₄
MDL Number:MFCD00007251
:167.12 g/mol
Merck Index:14,06588
Boiling Pt:341 °C (1013 hPa)
Melting Pt:139…142 °C
Density:1,494 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-508-5
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Melting point:141.0 to 144.0 deg-C
- Solubility in Methanol:almost transparency