Ethylvanillin ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:121-32-4
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00006944
:166.18 g/mol
Merck Index:14,03859
Boiling Pt:285 °C (1013 hPa)
Melting Pt:75…77 °C
Flash Pt:100 °C
EINECS:204-464-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:76.0 to 79.0 deg-C
- Solubility in Methanol:almost transparency