1-Bromo-4-nitrobenzene ≥99.0% (by GC)
: TCI
:586-78-7
:
:C₆H₄BrNO₂
:MFCD00007280
:202.01 g/mol
:255 °C (1013 hPa)
:124…126 °C
:1,93 g/cm³ (20 °C)
:≥ 110 °C
:Ambient
:209-583-8
:3459
:6.1,III
:GHS06
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:125.0 to 128.0 deg-C
- Solubility in hot EtOH:almost transparency