1-Bromo-4-chloro-3-fluorobenzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:60811-18-9
:
Formula:C₆H₃BrClF
MDL Number:MFCD00042519
:209.44 g/mol
Boiling Pt:142°C
Density:1,71 g/cm³ (20 °C)
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %