Ethyl-4-nitrocinnamate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:953-26-4
:
Formula:C₁₁H₁₁NO₄
MDL Number:MFCD00007380
:221.21 g/mol
EINECS:213-463-0
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:136.0 to 140.0 deg-C