Order Entry
Norway
ContactUsLinkComponent
Benzyl acetate ≥99.0% (by GC)
Irritant
Health
Benzyl acetate ≥99.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
Health
Benzyl acetate ≥99.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 99.0 %

Specification Test Results