
3-Methyl-2-pentanone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:565-61-7
:
Formula:C₆H₁₂O
MDL Number:MFCD00009336
:100.16 g/mol
Boiling Pt:116°C
Density:0,815 g/cm³ (20 °C)
Flash Pt:12°C
EINECS:209-282-1
UN:1224
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %