

2,6-Dimethyl-4-heptanone ≥90.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:108-83-8
:
Formula:C₉H₁₈O
MDL Number:MFCD00008940
:142.24 g/mol
Boiling Pt:168°C
Melting Pt:–46 °C
Density:0,806 g/cm³ (20 °C)
Flash Pt:50°C
Storage Temperature:Ambient
EINECS:203-620-1
UN:1157
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 90.0 %
Specification Test Results
Frequently Bought Together