
Potassium (4-cyanophenyl)trifluoroborate ≥95.0% (by total nitrogen basis)
TCIAP1815-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 95.0 %
:
:209.02 g/mol
:GHS07
:850623-36-8
:C₇H₄BF₃KN
:MFCD04115736