
Potassium (3-cyanophenyl)trifluoroborate ≥98.0% (by total nitrogen basis)
TCIAP1814-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
:
:209.02 g/mol
:180°C
:GHS07
:850623-46-0
:C₇H₄BF₃KN
:MFCD04115744