n-Heptadecane ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:629-78-7
:
Formula:H₃C(CH₂)₁₅CH₃
MDL Number:MFCD00009002
:240.47 g/mol
Boiling Pt:303°C
Melting Pt:21.2 °C
Density:0,7783 g/cm³ (20 °C)
Flash Pt:148 °C
EINECS:211-108-4
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White or Colorless powder to lump to clear liquid
- Purity (GC):min. 99.0 %