Palmitic acid ≥99.5% (by GC)
TCIAP1145-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.5 %
- Purity (Neutralization titration):min. 99.0 %
- Melting point:62.0 to 66.0 deg-C
- Solubility in hot Methanol:almost transparency
:
:14,06996
:352°C
:61…63 °C
:0,853 g/cm³ (20 °C)
:256.43 g/mol
:Ambient
:200-312-9
:GHS07
:57-10-3
:C₁₆H₃₂O₂
:MFCD00002747