Order Entry
Norway
ContactUsLinkComponent
1,1-Dimethoxyacetone ≥97.0% (by GC)
Flammable
Irritant
1,1-Dimethoxyacetone ≥97.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
Irritant
1,1-Dimethoxyacetone ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Red to Green clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

Frequently Bought Together