Order Entry
Norway
ContactUsLinkComponent
 
 
(±)-2,2-Dimethyl-3-hexanol ≥98.0% (by GC)
  TCIAD1532-10ML
 :  TCI
 :  
undefined
 
(±)-2,2-Dimethyl-3-hexanol ≥98.0% (by GC)
  TCIAD1532-10ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :130.23 g/mol
 :156°C
 :224-135-1
 :1987
 :3,III
 :GHS02
 :4209-90-9
 :C₈H₁₈O
 :MFCD00021929