
2-Bromo-5-(trifluoromethoxy)phenol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:205371-26-2
:
Formula:C₇H₄BrF₃O₂
MDL Number:MFCD06660162
:257.01 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together