
Sodium diphenylamine-4-sulphonate ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:6152-67-6
:
Formula:C₁₂H₁₀NNaO₃S
MDL Number:MFCD00036468
:271.27 g/mol
Merck Index:13,07397
EINECS:228-165-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Light gray to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Solubility in Water:almost transparency
- Sensitiveness:to pass test