

1,2,4-Triazole ≥99.0% (by GC)
: TCI
:288-88-0
:
:C₂H₃N₃
:MFCD00005228
:69.07 g/mol
:14,09605
:260°C
:116…119 °C
:1,394 g/cm³ (25 °C)
:206-022-9
:GHS07
:GHS08
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:119.0 to 123.0 deg-C
- Solubility in Methanol:almost transparency