Order Entry
Norway
ContactUsLinkComponent
Nuclear Fast Red
Irritant
Nuclear Fast Red
Supplier:  TCI
Certificates
undefined
Irritant
Nuclear Fast Red
Supplier:  TCI
Specification Test Results
  • Appearance:
    Dark red to Brown powder to crystal
  • Lambda max.:
    532.0 to 536.0 nm (0.1mol/L NaOH sol.)
  • Absorbance (E1%1cm):
    min. 380 (0.1mol/L NaOH sol., 532.0 to 536.0 nm)