Order Entry
Norway
ContactUsLinkComponent
 
Eosin Y ≥90.0% (by UV/Vis)
Catalog # TCIAT0037-25G
Supplier:  TCI
CAS Number:  
undefined
Eosin Y ≥90.0% (by UV/Vis)
Catalog # TCIAT0037-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pakketype:
    Glass bottle
  • Fp:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Orange to Brown to Dark red powder to crystaline
  • Purity (UV-vis method):
    min. 90.0 % (calcd.on dried substance)
  • Lambda max.:
    515.0 to 519.0 nm (0.02mol/L AcONH4 sol.)
  • Drying loss:
    max. 15.0 %

Specification Test Results

About this item

 :
Merck Index:14,03603
Melting Pt:295…296 °C
 :691.86 g/mol
EINECS:241-409-6
 :GHS07
Storage Temperature:Romtemperatur
CAS Number:17372-87-1
Formula:C₂₀H₆Br₄Na₂O₅
MDL Number:MFCD00005040