Order Entry
Norway
ContactUsLinkComponent
2-Octynoic acid methyl ester ≥98.0% (by GC)
2-Octynoic acid methyl ester ≥98.0% (by GC)
Catalog # TCIAO0239-25ML
Supplier:  TCI
CAS Number:  
undefined
2-Octynoic acid methyl ester ≥98.0% (by GC)
Catalog # TCIAO0239-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:220°C
Flash Pt:100°C
 :154.21 g/mol
EINECS:203-836-6
 :GHS07
CAS Number:111-12-6
Formula:C₉H₁₄O₂
MDL Number:MFCD00009530