Order Entry
Norway
ContactUsLinkComponent
 
2-Nonynoic acid methyl ester ≥98.0% (by GC)
  TCIAN0595-25ML
 :  TCI
 :  
undefined
2-Nonynoic acid methyl ester ≥98.0% (by GC)
  TCIAN0595-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :168.24 g/mol
 :121°C
 :104°C
 :203-909-2
 :111-80-8
 :C₁₀H₁₆O₂
 :MFCD00009547