Order Entry
Norway
ContactUsLinkComponent
4-Amino-m-cresol ≥98.0% (by GC, titration analysis)
4-Amino-m-cresol ≥98.0% (by GC, titration analysis)
  TCIAA0288-25G
 :  TCI
 :  
undefined
4-Amino-m-cresol ≥98.0% (by GC, titration analysis)
  TCIAA0288-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :123.15 g/mol
 :259 °C (1013 hPa)
 :177…179 °C
 :220-621-2
 :3077
 :9,III
 :GHS07
 :GHS08
 :GHS09
 :2835-99-6
 :C₇H₉NO
 :MFCD00007871