Order Entry
Norway
ContactUsLinkComponent
3-Thiophenemethanol ≥96.0% (by GC)
3-Thiophenemethanol ≥96.0% (by GC)
Supplier:  TCI
Certificates
undefined
3-Thiophenemethanol ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results