Squalene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:111-02-4
:
Formula:C₃₀H₅₀
MDL Number:MFCD00008912
:410.73 g/mol
Merck Index:13,08847
Boiling Pt:252°C
Melting Pt:–5°C
Density:0,86 g/cm³ (20 °C)
Flash Pt:220°C
EINECS:203-826-1
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %