

1,4-Dimethylpiperazine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:106-58-1
:
Formula:C₆H₁₄N₂
MDL Number:MFCD00006156
:114.19 g/mol
Boiling Pt:133°C
Melting Pt:–1 °C
Density:0,85 g/cm³ (20 °C)
Flash Pt:25°C
EINECS:203-412-0
UN:2924
ADR:3,III
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %