
2,5-Dichloro-p-phenylenediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:20103-09-7
:
Formula:C₆H₆Cl₂N₂
MDL Number:MFCD00007902
:177.03 g/mol
EINECS:243-512-1
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:164.0 to 168.0 deg-C