Order Entry
Norway
ContactUsLinkComponent
 
 
N,N'-Diphenyl-p-phenylenediamine ≥95.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
N,N'-Diphenyl-p-phenylenediamine ≥95.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Gray to Brown powder to lump
  • Purity (GC):
    min. 90.0 %
  • Purity (Nonaqueous Titration):
    min. 95.0 %