Order Entry
Norway
ContactUsLinkComponent
Ethyl m-tolylacetate ≥98.0% (by GC)
Ethyl m-tolylacetate ≥98.0% (by GC)
Catalog # TCIAT1226-10G
Supplier:  TCI
CAS Number:  
undefined
Ethyl m-tolylacetate ≥98.0% (by GC)
Catalog # TCIAT1226-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :178.23 g/mol
Boiling Pt:238°C
Density:1,018 g/cm³ (17.5 °C)
EINECS:254-778-3
CAS Number:40061-55-0
Formula:(C₆H₄(CH₃))CH₂COOCH₂CH₃
MDL Number:MFCD00017273