Order Entry
Norway
ContactUsLinkComponent
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Supplier:  TCI
Certificates
undefined
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity:
    min. 98.0 area%
  • Optical purity:
    min. 99.0 ee%

Specification Test Results

Related Information

Frequently Bought Together