Order Entry
Norway
ContactUsLinkComponent
 
cis-4-Hydroxy-D(+)-proline ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
cis-4-Hydroxy-D(+)-proline ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +56.0 to +60.0 deg (C=2, H2O)