
meso-2,3-Dibromosuccinic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:608-36-6
:
Formula:HOOC(BrCH)₂COOH
MDL Number:MFCD00066439
Merck Index:14,03029
Boiling Pt:352 °C (1013 hPa)
Melting Pt:272 °C
EINECS:452-290-7
UN:3261
ADR:8,III
:275.88 g/mol
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency
- Solubility in hot Water:almost transparency