

(±)-2,3-Dibromopropionic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:600-05-5
:
Formula:BrH₂CBrCHCOOH
MDL Number:MFCD00004212
:231.87 g/mol
Boiling Pt:119°C
Melting Pt:65 °C
Storage Temperature:Refrigerator
EINECS:209-981-1
UN:3261
ADR:8,III
:GHS05
:GHS08
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):min. 97.0 %
- Melting point:61.0 to 68.0 deg-C
- Solubility in Water:very faint turbidity
- Bromide:max. 0.2 %