
3-Hydroxy-2-nitropyridine ≥98.0% (by GC, titration analysis)
TCIAH0764-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:69.0 to 73.0 deg-C
- Solubility in Methanol:almost transparency
:
:140.1 g/mol
:70 °C
:239-191-2
:GHS07
:15128-82-2
:C₅H₄N₂O₃
:MFCD00006258