Order Entry
Norway
ContactUsLinkComponent
Dansyl fluoride ≥98.0% (by HPLC, titration analysis)
Corrosive
Dansyl fluoride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD2197-1G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Dansyl fluoride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD2197-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    50.0 to 53.0 deg-C
  • Solubility in Acetone:
    almost transparency

Specification Test Results

About this item

 :
 :253.3 g/mol
EINECS:252-071-4
UN:3261
ADR:8,III
 :GHS05
CAS Number:34523-28-9
Formula:C₁₂H₁₂FNO₂S
MDL Number:MFCD00042702