
1,5-Bis(diphenylphosphino)pentane ≥96.0% (by GC, titration analysis)
Catalog # TCIAB1960-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystalline
- Purity (GC):min. 96.0 %
- Purity (Iodometric Titration):min. 96.0 %
- Melting point:43.0 to 46.0 deg-C
- Solubility in N,N-DMF:almost transparency
Specification Test Results
About this item
:
:440.5 g/mol
Boiling Pt:230 °C (0,1 -rr)
Melting Pt:43…46 °C
:GHS07
CAS Number:27721-02-4
Formula:C₂₉H₃₀P₂
MDL Number:MFCD00003052