Order Entry
Norway
ContactUsLinkComponent
 
 
2-Methoxy-4-nitrophenol ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
2-Methoxy-4-nitrophenol ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    101.0 to 105.0 deg-C