Order Entry
Norway
ContactUsLinkComponent
α-Linolenic acid ≥70% (by GC)
α-Linolenic acid ≥70% (by GC)
  TCIAL0050-25ML
 :  TCI
 :  
undefined
α-Linolenic acid ≥70% (by GC)
  TCIAL0050-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 70.0 %

 

 

 :
 :278.44 g/mol
 :14,05506
 :232°C
 :-11°C
 :–11°C
 :0,9141 g/cm³ (18 °C)
 :Refrigerator
 :207-334-8
 :GHS07
 :463-40-1
 :C₁₈H₃₀O₂
 :MFCD00065720