Dibenzyl malonate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:15014-25-2
:
Formula:C₁₇H₁₆O₄
MDL Number:MFCD00004779
:284.31 g/mol
Boiling Pt:234°C
Density:1,137 g/cm³ (20 °C)
EINECS:239-099-2
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %