Dimethyl glyoxime ≥98.0% (W)
Supplier: TCI
Certificates
About this item
CAS Number:95-45-4
:
Formula:CH₃C(=NOH)C(=NOH)CH₃
MDL Number:MFCD00002117
:116.12 g/mol
Merck Index:14,03246
Boiling Pt:53 °C (8 -rr)
Melting Pt:240°C
Density:1,37 g/cm³ (20 °C)
EINECS:202-420-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Gravimetric Method):min. 98.0 %
- Solubility in EtOH:almost transparency
- Sensitiveness:Colorimetric Test : Turn Yellow with Nickel