
2-Hydroxy-9-fluorenone ≥90.0% (by HPLC, titration analysis)
TCIAH0329-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:Light yellow to Brown powder to crystaline
- Purity (HPLC):min. 90.0 area%
- Purity (Neutralization titration):min. 90.0 %
- Melting point:204.0 to 210.0 deg-C
:
:196.21 g/mol
:353 °C (1013 hPa)
:213.5…214.5 °C
:230-119-5
:GHS07
:6949-73-1
:C₁₃H₈O₂
:MFCD00001155