Order Entry
Norway
ContactUsLinkComponent
 
3,5-Bis(trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)
 :  TCI
 
undefined
3,5-Bis(trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Red to Green clear liquid
  • Purity (GC):
    min. 98.0 %