4-Nitrobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:98-74-8
:
Formula:C₆H₄ClNO₄S
MDL Number:MFCD00007445
:221.62 g/mol
Boiling Pt:181°C
Melting Pt:79 °C
Density:1,69 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-697-9
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:77.0 to 80.0 deg-C
- Solubility in Toluene:within almost transparency