
2,6-Difluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
: TCI
:60230-36-6
:
:C₆H₃ClF₂O₂S
:MFCD00085004
:212.6 g/mol
:129°C
:1,568 g/cm³ (20 °C)
:3265
:8,III
:GHS05
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %