Order Entry
Norway
ContactUsLinkComponent
2'-Hydroxyacetanilide ≥99.0% (by HPLC, total nitrogen)
2'-Hydroxyacetanilide ≥99.0% (by HPLC, total nitrogen)
Catalog # TCIAH0189-25G
Supplier:  TCI
CAS Number:  
2'-Hydroxyacetanilide ≥99.0% (by HPLC, total nitrogen)
Catalog # TCIAH0189-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Purity (HPLC):
    min. 99.0 area%
  • Purity (with Total Nitrogen):
    min. 99.0 %
  • Melting point:
    208.0 to 212.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :151.16 g/mol
EINECS:210-396-9
 :GHS07
CAS Number:614-80-2
Formula:C₈H₉NO₂
MDL Number:MFCD00002181