L(+)-Valine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:72-18-4
:
Formula:C₅H₁₁NO₂
MDL Number:MFCD00064220
:117.15 g/mol
Merck Index:14,09909
Boiling Pt:217 °C (1013 hPa)
Melting Pt:86 °C
Density:1,284 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-773-6
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Optical purity (LC):min. 99.0 ee%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency