(±)-trans-1,2-Cyclohexanediol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1460-57-7
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00063611
Boiling Pt:120°C
Melting Pt:100…103 °C
Density:1,147 g/cm³ (24 °C)
:116.16 g/mol
EINECS:215-956-6
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:102.0 to 105.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together