Order Entry
Norway
ContactUsLinkComponent
 
(±)-1-Hexyn-3-ol ≥95.0% (by GC)
 :  TCI
 
undefined
(±)-1-Hexyn-3-ol ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 95.0 %