
Fumaric acid ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:110-17-8
:
Formula:C₄H₄O₄
MDL Number:MFCD00002700
:116.07 g/mol
Merck Index:14,04287
Boiling Pt:356 °C (1013 hPa)
Melting Pt:287°C
Density:1,625 g/cm³ (20 °C)
Flash Pt:273 °C
EINECS:203-743-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Solubility in Methanol:almost transparency