Order Entry
Norway
ContactUsLinkComponent
 
Chlorocycloheptane ≥98.0% (by GC)
  TCIAC0681-10ML
 :  TCI
 :  
undefined
Chlorocycloheptane ≥98.0% (by GC)
  TCIAC0681-10ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :132.63 g/mol
 :175°C
 :1,0133 g/cm³ (0 °C)
 :219-522-7
 :1993
 :3,III
 :GHS02
 :2453-46-5
 :C₇H₁₃Cl
 :MFCD00046501